Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Metrnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144797.3 | NP_659046.1 | 4 | 1318 | ||
GenBank mRNA | AK034855.1 | - | 4 | 1319 | ||
AK085880.1 | - | 4 | 1320 | |||
AK170549.1 | - | 3 | 933 | |||
AK170800.1 | - | 4 | 1320 | |||
AK171200.1 | - | 4 | 1064 | |||
BC019776.1 | - | 4 | 1273 | 3672224 | ||
BC024445.1 | - | 4 | 1333 | 4974030 | ||
BC024497.1 | - | 1321 | 4982563 | |||
BC026646.1 | - | 4 | 1312 | 4987595 |