Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm6sf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291282.1 | NP_001278211.1 | 6 | 469 | ||
NM_145375.4 | NP_663350.2 | 6 | 977 | |||
GenBank mRNA | AK046714.1 | - | 6 | 878 | ||
AK083132.1 | - | 6 | 980 | |||
AK135100.1 | - | 6 | 959 | |||
AK136581.1 | - | 6 | 470 | |||
AK162572.1 | - | 5 | 746 | |||
AK163621.1 | - | 6 | 902 | |||
AK172109.1 | - | 6 | 893 | |||
AK172145.1 | - | 6 | 878 | |||
AY512939.1 | - | 2 | 1094 | |||
BC023123.1 | - | 6 | 859 | 3995280 |