Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029790.3 | NP_084066.2 | 4 | 568 | ||
GenBank mRNA | AK002682.1 | - | 4 | 569 | ||
BC089533.1 | - | 5 | 695 | |||
BC106186.1 | - | 4 | 540 | 30244889 | ||
BC115926.1 | - | 4 | 527 | 40105262 | ||
BC117549.1 | - | 4 | 527 | 40105259 |