Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tchp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029992.2 | NP_084268.2 | 4, 5 | 643 | ||
GenBank mRNA | AK020912.1 | - | 4, 5 | 606 | ||
AK083027.1 | - | 4, 5 | 559 | |||
AK159947.1 | - | 4, 5 | 604 | |||
BC043048.1 | - | 4, 5 | 548 | 5704036 | ||
BU512279.1 | - | 4, 5 | 643 |