Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030024.2 | NP_084300.1 | 2 | 728 | ||
GenBank mRNA | AJ132433.2 | - | 2 | 728 | ||
AK021401.1 | - | 2 | 836 | |||
AK136438.1 | - | 2 | 908 | |||
BC055838.1 | - | 2 | 857 | |||
BC132416.1 | - | 1 | 404 | 40130693 | ||
BC138757.1 | - | 1 | 404 | 8861779 |