Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aff4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033565.2 | NP_291043.1 | 2, 3 | 486 | ||
GenBank mRNA | AF190449.1 | - | 2, 3 | 428 | ||
AK044058.1 | - | 2, 3 | 370 | |||
AK054401.1 | - | 2, 3 | 334 | |||
BC099959.1 | - | 1, 2 | 153 | |||
BC138999.1 | - | 2, 3 | 159 | 8862021 | ||
BC145490.1 | - | 2, 3 | 159 | 9054053 |