Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekho2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153119.3 | NP_694759.1 | 6 | 1943 | ||
GenBank mRNA | AK154092.1 | - | 6 | 1838 | ||
AK158288.1 | - | 6 | 1911 | |||
AK170479.1 | - | 6 | 1927 | |||
AK184864.1 | - | 1 | 392 | |||
BC025598.1 | - | 1 | 820 | |||
BC028908.1 | - | 6 | 1902 | 3668775 |