Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttyh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK052340.1 | - | 12 | 1516 | ||
AK173233.1 | - | 9 | 905 | |||
AK183225.1 | - | 2 | 199 | |||
BC059083.1 | - | 12 | 1516 | 6827289 | ||
BC062917.1 | - | 12 | 1522 | 6831835 | ||
CV784391.1 | - | 1 | 60 |