Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4632415L05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014580.1 | - | 1 | 934 | ||
BC023403.1 | - | 1 | 956 | 5252327 | ||
BC037267.1 | - | 1 | 944 |