Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027891.4 | NP_082167.2 | 12 | 1619 | ||
GenBank mRNA | AK004717.1 | - | 10 | 2581 | ||
AK084948.1 | - | 12 | 1619 | |||
AK085327.1 | - | 1 | 1650 | |||
AK153976.1 | - | 12 | 1590 | |||
AK154352.1 | - | 12 | 1561 | |||
AK167020.1 | - | 12 | 1568 | |||
AK172708.1 | - | 12 | 1587 | |||
AK188535.1 | - | 2 | 78 | |||
AK207344.1 | - | 4 | 326 | |||
BC048397.1 | - | 12 | 1598 | 6396815 |