Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028679.3 | NP_082955.2 | 2 | 375 | ||
GenBank mRNA | AF461763.1 | - | 2 | 355 | ||
AJ440757.2 | - | 2 | 297 | |||
AK014783.1 | - | 2 | 426 | |||
AK028436.1 | - | 288 | ||||
AK029057.1 | - | 2 | 376 | |||
AK049210.1 | - | 2 | 406 | |||
BC120829.1 | - | 2 | 336 | 40129752 | ||
BC120831.1 | - | 2 | 336 | 40129754 | ||
BC144835.1 | - | 2 | 336 |