Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163579.1 | NP_001157051.1 | 2 | 337 | ||
NM_001163580.1 | NP_001157052.1 | 2 | 337 | |||
NM_028915.3 | NP_083191.2 | 2 | 337 | |||
GenBank mRNA | AK004646.1 | - | 2 | 338 | ||
AK029563.1 | - | 2 | 254 | |||
AK030355.1 | - | 1 | 2255 | |||
AK173256.1 | - | 2 | 265 | |||
BC061202.1 | - | 2 | 293 | |||
BC099680.1 | - | 2 | 301 | |||
BC138734.1 | - | 2 | 311 | 8861756 | ||
BC138735.1 | - | 2 | 311 | 8861757 |