Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029402.3 | NP_083678.2 | 2 | 296 | ||
GenBank mRNA | AK004875.2 | - | 2 | 296 | ||
AK016520.1 | - | 2 | 266 | |||
AK028589.1 | - | 2 | 265 | |||
AK144693.1 | - | 2 | 269 | |||
AK160597.1 | - | 2 | 281 | |||
AK220287.1 | - | 2 | 271 | |||
BC026779.1 | - | 2 | 197 | 4018298 | ||
BC027428.1 | - | 2 | 259 | 3983461 |