Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap5m1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144535.4 | NP_653118.3 | 1, 2 | 380 | ||
GenBank mRNA | AK016540.1 | - | 1, 2 | 327 | ||
AK028927.1 | - | 1, 2 | 333 | |||
AK030928.1 | - | 1, 2 | 307 | |||
AK031527.1 | - | 1, 2 | 779 | |||
AK036454.1 | - | 1, 2 | 305 | |||
AK036465.1 | - | 1, 2 | 312 | |||
AK041400.1 | - | 1, 2 | 333 | |||
AK042544.1 | - | 1, 2 | 302 | |||
AK051372.1 | - | 1, 2 | 327 | |||
AK154166.1 | - | 1, 2 | 333 | |||
AK164148.1 | - | 1, 2 | 151 | |||
AK170232.1 | - | 1, 2 | 157 | |||
AK211895.1 | - | 1, 2 | 57 | |||
BC023094.1 | - | 1, 2 | 380 | 4017596 |