Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173402.2 | NP_775578.2 | 2 | 1213 | ||
GenBank mRNA | AK053804.1 | - | 2 | 1214 | ||
AK144822.1 | - | 2 | 1195 | |||
AK145890.1 | - | 2 | 1163 | |||
BC040396.1 | - | 2 | 1210 | 4461061 |