Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam189a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183087.4 | NP_898910.2 | 10 | 1610 | ||
GenBank mRNA | AK017751.1 | - | 4 | 716 | ||
AK138167.1 | - | 8 | 1106 | |||
AK172974.1 | - | 10 | 1382 | |||
BC049085.1 | - | 7 | 932 | |||
BC147145.1 | - | 2 | 324 | |||
BC147146.1 | - | 2 | 324 |