Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4f16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024442.1 | NP_077762.1 | 13 | 2140 | ||
GenBank mRNA | AF233646.1 | - | 13 | 2140 | ||
AK078557.1 | - | 13 | 2201 | |||
AK083886.1 | - | 12 | 2050 | |||
AK146232.1 | - | 13 | 2213 | |||
BC026539.1 | - | 13 | 2172 | 4982322 |