Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmtm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026036.3 | NP_080312.1 | 4 | 2677 | ||
GenBank mRNA | AK003445.1 | - | 1 | 574 | ||
AK012941.1 | - | 4 | 2679 | |||
AK030885.1 | - | 4 | 2657 | |||
AK128960.1 | - | 4 | 2658 | |||
AK145743.1 | - | 4 | 2658 | |||
AK150775.1 | - | 4 | 2676 | |||
AK151092.1 | - | 4 | 2676 | |||
AK151249.1 | - | 1 | 2196 | |||
AK151431.1 | - | 4 | 2676 | |||
AK152448.1 | - | 4 | 2679 | |||
AK152723.1 | - | 4 | 2688 | |||
AK152817.1 | - | 4 | 2688 | |||
AK153092.1 | - | 4 | 2676 | |||
AK154481.1 | - | 4 | 2665 | |||
AK159439.1 | - | 4 | 2647 | |||
AK159735.1 | - | 4 | 2672 | |||
AK165948.1 | - | 4 | 2646 | |||
AK167499.1 | - | 4 | 2664 | |||
AK167854.1 | - | 4 | 2669 | |||
AK170905.1 | - | 4 | 2660 | |||
AK213137.1 | - | 1 | 19 | |||
AY241868.1 | - | 4 | 2714 | |||
BC007194.1 | - | 2 | 2320 | |||
BC027248.1 | - | 4 | 2649 | 2811938 |