Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026384.3 | NP_080660.1 | 8 | 1565 | ||
GenBank mRNA | AK002443.1 | - | 8 | 1571 | ||
BC043447.1 | - | 8 | 1565 | 5125951 |