Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B230118H07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026592.3 | NP_080868.1 | 4, 5 | 496 | ||
GenBank mRNA | AK010820.1 | - | 4, 5 | 459 | ||
AK020980.1 | - | 4, 5 | 497 | |||
AK051679.1 | - | 4, 5 | 497 | |||
AK088637.1 | - | 4, 5 | 540 | |||
BC025075.1 | - | 4, 5 | 500 | 4037434 | ||
DQ112091.1 | - | 709 | ||||
DQ112092.1 | - | 501 |