Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026929.4 | NP_081205.1 | 3 | 1533 | ||
GenBank mRNA | AK007378.1 | - | 3 | 1485 | ||
AK032218.1 | - | 3 | 1521 | |||
BC025169.1 | - | 3 | 1474 | 4483043 |