Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eqtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290623.1 | NP_001277552.1 | 9 | 979 | ||
NM_027089.4 | NP_081365.2 | 8 | 1102 | |||
GenBank mRNA | AK006446.1 | - | 8 | 1053 | ||
AK016738.1 | - | 9 | 950 | |||
BC089517.1 | - | 8 | 1069 | 6740450 | ||
DQ336137.1 | - | 9 | 930 | |||
EU828781.1 | - | 8 | 1087 | |||
EU828782.1 | - | 9 | 964 |