Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027194.3 | NP_081470.2 | 4 | 1240 | ||
GenBank mRNA | AK010545.1 | - | 4 | 1172 | ||
AK045768.1 | - | 4 | 1199 | |||
AK080854.1 | - | 4 | 1167 | |||
AK083654.1 | - | 4 | 1161 | |||
AK085975.1 | - | 4 | 1193 | |||
AK086015.1 | - | 4 | 1176 | |||
AK086059.1 | - | 4 | 1195 | |||
AK088246.1 | - | 4 | 1704 | |||
AK149407.1 | - | 4 | 1169 | |||
AK160039.1 | - | 4 | 1177 | |||
AK166012.1 | - | 4 | 1193 | |||
AK197347.1 | - | 1 | 187 | |||
BC026789.1 | - | 4 | 1167 | 4188168 |