Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyttd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159349.1 | NP_001152821.1 | 8 | 1592 | ||
NM_027226.4 | NP_081502.2 | 9 | 1694 | |||
GenBank mRNA | AK014447.1 | - | 9 | 1666 | ||
AK029785.1 | - | 9 | 1640 | |||
AK050448.1 | - | 9 | 1653 | |||
AK077184.1 | - | 10 | 1602 | |||
AK139638.1 | - | 8 | 1588 | |||
AK161252.1 | - | 9 | 1672 | |||
AK165633.1 | - | 9 | 1650 | |||
AK166032.1 | - | 9 | 1666 | |||
AK167564.1 | - | 1 | 710 | |||
AK170511.1 | - | 9 | 1666 | |||
BC028253.1 | - | 9 | 1646 | 3983533 |