Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025105.2 | NP_001020276.1 | 1, 2 | 719 | ||
NM_001271741.1 | NP_001258670.1 | 1, 2 | 719 | |||
NM_001892.5 | NP_001883.4 | 1, 2 | 719 | |||
GenBank mRNA | AF218004.1 | - | 1, 2 | 517 | ||
AK294942.1 | - | 2, 3 | 306 | |||
AK303794.1 | - | 2, 3 | 352 | |||
BC007246.1 | - | 1, 2 | 540 | |||
BC008717.2 | - | 1, 2 | 261 | 2822250 | ||
BC021971.2 | - | 1, 2 | 538 | |||
BC025371.2 | - | 1, 2 | 538 | |||
BT019829.1 | - | 1, 2 | 131 | |||
BT019830.1 | - | 1, 2 | 131 | |||
CR542206.1 | - | 1, 2 | 131 | |||
DQ082865.1 | - | 1, 2 | 131 | |||
EU668329.1 | - | 1, 2 | 131 | |||
L37042.1 | - | 1, 2 | 342 |