Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281303.1 | NP_001268232.1 | 12, 13 | 1247 | ||
NM_006415.3 | NP_006406.1 | 12, 13 | 1247 | |||
XM_011518139.2 | XP_011516441.1 | 12, 13 | 1231 | |||
XM_017014200.1 | XP_016869689.1 | 13, 14 | 1292 | |||
XM_017014201.1 | XP_016869690.1 | 13, 14 | 1364 | |||
XM_017014202.1 | XP_016869691.1 | 8, 9 | 1425 | |||
GenBank mRNA | AK291546.1 | - | 12, 13 | 1147 | ||
AK309775.1 | - | 1019 | ||||
BC068537.1 | - | 12, 13 | 1151 | 5259635 | ||
Y08685.1 | - | 12, 13 | 1143 |