Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294308.1 | NP_001281237.1 | 4 | 985 | ||
NM_022414.2 | NP_071859.1 | 4 | 973 | |||
GenBank mRNA | AK140173.1 | - | 4 | 969 | ||
AK165775.1 | - | 4 | 779 | |||
BC024263.1 | - | 4 | 973 | 5133442 |