Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104530.1 | NP_001098000.1 | 3 | 380 | ||
NM_001104556.1 | NP_001098026.1 | 2 | 378 | |||
NM_001104557.1 | NP_001098027.1 | 2 | 210 | |||
NM_025426.3 | NP_079702.3 | 3 | 548 | |||
GenBank mRNA | AK004354.1 | - | 3 | 513 | ||
AK010357.1 | - | 3 | 376 | |||
AK012799.1 | - | 3 | 528 | |||
AK144918.1 | - | 2 | 336 | |||
AK147739.1 | - | 3 | 538 | |||
AK167407.1 | - | 3 | 508 | |||
BC011333.1 | - | 3 | 535 | 3993084 | ||
BE333513.1 | - | 2 | 308 | |||
CK030901.1 | - | 3 | 380 | |||
CK788351.1 | - | 200 |