Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nicn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025449.3 | NP_079725.1 | 2 | 258 | ||
GenBank mRNA | AJ299741.1 | - | 2 | 250 | ||
AK002789.1 | - | 2 | 264 | |||
AK013602.1 | - | 2 | 286 | |||
AK075732.1 | - | 2 | 266 | |||
AK146890.1 | - | 2 | 268 | |||
AK169135.1 | - | 2 | 268 | |||
BC024050.1 | - | 2 | 258 | 5133763 |