Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025458.2 | NP_079734.1 | 3 | 410 | ||
GenBank mRNA | AK007320.1 | - | 3 | 406 | ||
AK007518.1 | - | 3 | 411 | |||
AK007830.1 | - | 3 | 404 | |||
AK007885.1 | - | 3 | 411 | |||
BC086923.1 | - | 3 | 410 | 6760985 |