Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320412.1 | NP_001307341.1 | 6 | 751 | ||
NM_006657.2 | NP_006648.1 | 6 | 751 | |||
NM_206965.1 | NP_996848.1 | 6 | 751 | |||
XM_006723961.3 | XP_006724024.2 | 6 | 1042 | |||
XM_006723962.3 | XP_006724025.2 | 6 | 1650 | |||
XM_011529434.2 | XP_011527736.1 | 6 | 1665 | |||
XM_011529435.2 | XP_011527737.1 | 6 | 1049 | |||
XM_011529436.2 | XP_011527738.1 | 6 | 1650 | |||
XM_011529437.2 | XP_011527739.1 | 6 | 1044 | |||
XM_011529439.2 | XP_011527741.1 | 4 | 454 | |||
XM_011529440.2 | XP_011527742.1 | 6 | 1036 | |||
GenBank mRNA | AF169017.1 | - | 6 | 751 | ||
AF289021.1 | - | 6 | 751 | |||
AF289022.1 | - | 6 | 751 | |||
AF289023.1 | - | 6 | 751 | |||
AF289024.1 | - | 6 | 776 | |||
BC052248.1 | - | 6 | 747 | 6270063 | ||
BC136383.1 | - | 6 | 743 | 9020370 | ||
BC136395.1 | - | 6 | 743 | 9020382 |