Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf19a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013923.2 | NP_038951.1 | 10 | 2369 | ||
GenBank mRNA | AF120206.1 | - | 2262 | |||
AF120207.1 | - | 9 | 2262 | |||
AK082665.1 | - | 8 | 1963 | |||
AK147768.1 | - | 10 | 2357 | |||
BC040769.1 | - | 9 | 2144 | 4457807 | ||
X71642.1 | - | 6 | 916 |