Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016699.2 | NP_057908.2 | 1, 2 | 150 | ||
GenBank mRNA | AF091392.1 | - | 1, 2 | 119 | ||
AK017621.1 | - | 1, 2 | 123 | |||
AK081014.1 | - | 1, 2 | 146 | |||
AK144162.1 | - | 1, 2 | 135 | |||
AK162887.1 | - | 1, 2 | 119 | |||
AK166070.1 | - | 1, 2 | 134 | |||
AK168700.1 | - | 1, 2 | 123 | |||
BC027326.1 | - | 1, 2 | 128 | 3987812 | ||
BC027840.1 | - | 1, 2 | 138 | 5325071 | ||
BY277385.1 | - | 1, 2 | 158 |