Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016710.2 | NP_057919.2 | 6 | 1023 | ||
GenBank mRNA | AB018374.2 | - | 6 | 978 | ||
AF213454.1 | - | 6 | 1003 | |||
AK131996.1 | - | 6 | 993 | |||
AK146755.1 | - | 6 | 1000 | |||
BC021626.1 | - | 6 | 975 | 4219308 | ||
BC083087.1 | - | 6 | 985 | 6518798 |