Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006713.3 | NP_006704.3 | 5 | 871 | ||
XM_011513944.2 | XP_011512246.1 | 6 | 1124 | |||
XM_017008986.1 | XP_016864475.1 | 6 | 967 | |||
XM_017008987.1 | XP_016864476.1 | 6 | 844 | |||
GenBank mRNA | AB209285.1 | - | 5 | 766 | ||
BC009610.1 | - | 5 | 799 | 4297490 | ||
BC010537.1 | - | 5 | 774 | 3457167 | ||
BC022339.1 | - | 5 | 804 | 4134117 | ||
BC063402.1 | - | 6 | 1089 | |||
BC071655.1 | - | 5 | 788 |