Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtmr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016985.2 | NP_058681.1 | 15 | 3439 | ||
GenBank mRNA | AK048714.1 | - | 7 | 2692 | ||
AK051390.1 | - | 8 | 2437 | |||
BC056376.1 | - | 15 | 3439 | 6849757 | ||
BC057337.1 | - | 15 | 3433 | 6825916 |