Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253700.1 | NP_001240629.1 | 7 | 887 | ||
NM_018740.2 | NP_061210.2 | 9 | 1369 | |||
GenBank mRNA | AF004107.1 | - | 8 | 1288 | ||
AK011130.1 | - | 7 | 1181 | |||
AK013662.1 | - | 7 | 970 | |||
AK152582.1 | - | 9 | 1373 | |||
AK207822.1 | - | 1 | 218 | |||
AK215451.1 | - | 2 | 285 | |||
BC003752.1 | - | 8 | 1220 | 3489878 |