Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018859.2 | NP_061347.2 | 6 | 586 | ||
GenBank mRNA | AK002507.1 | - | 6 | 561 | ||
AK007924.1 | - | 6 | 563 | |||
AK028605.1 | - | 412 | ||||
BC012692.1 | - | 6 | 602 | 3986863 | ||
U68535.1 | - | 6 | 572 |