Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smad9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019483.5 | NP_062356.3 | 5 | 1442 | ||
GenBank mRNA | AF175408.2 | - | 4 | 1120 | ||
AK137411.1 | - | 5 | 1443 | |||
AY145520.1 | - | 4 | 1275 | |||
BC119141.1 | - | 4 | 1159 | 8733890 |