Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019657.4 | NP_062631.1 | 4 | 356 | ||
GenBank mRNA | AF064635.1 | - | 4 | 335 | ||
AK078841.1 | - | 4 | 357 | |||
AK081869.1 | - | 4 | 350 | |||
AK082715.1 | - | 4 | 350 | |||
AK088521.1 | - | 4 | 350 | |||
AK150849.1 | - | 4 | 351 | |||
AK152489.1 | - | 4 | 351 | |||
AK166027.1 | - | 4 | 350 | |||
AK167908.1 | - | 4 | 350 | |||
BC010749.1 | - | 4 | 332 | |||
BC037620.1 | - | 3 | 273 | 5375831 | ||
BC090659.1 | - | 4 | 326 | 30675153 |