Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019674.3 | NP_062648.1 | 8 | 836 | ||
GenBank mRNA | AF088911.1 | - | 8 | 861 | ||
AK027923.1 | - | 6 | 616 | |||
AK144244.1 | - | 7 | 763 | |||
AK199030.1 | - | 1 | 63 | |||
BC001993.1 | - | 8 | 836 | 3491568 | ||
U79747.1 | - | 4 | 488 |