Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Umodl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177465.4 | NP_803416.2 | 15 | 2957 | ||
GenBank mRNA | AK040446.1 | - | 7 | 1458 | ||
AY771616.1 | - | 16 | 2874 | |||
AY771617.1 | - | 15 | 2961 | |||
AY771618.1 | - | 15 | 2616 | |||
BC051427.1 | - | 15 | 2612 | |||
BC132381.1 | - | 2 | 146 |