Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Umodl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177465.4 | NP_803416.2 | 20 | 3896 | ||
GenBank mRNA | AK040446.1 | - | 12 | 2397 | ||
AY771616.1 | - | 21 | 3813 | |||
AY771617.1 | - | 20 | 3900 | |||
AY771618.1 | - | 20 | 3555 | |||
BC051427.1 | - | 20 | 3551 | |||
BC132381.1 | - | 7 | 1085 |