Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083328.1 | NP_001076797.1 | 7 | 784 | ||
NM_001083329.1 | NP_001076798.1 | 6 | 690 | |||
NM_028422.1 | NP_082698.1 | 7 | 829 | |||
NM_181424.3 | NP_852089.2 | 8 | 923 | |||
GenBank mRNA | AK040689.1 | - | 7 | 1313 | ||
BC025144.1 | - | 8 | 922 | 4483063 | ||
BC028465.1 | - | 2 | 225 | |||
BC030918.1 | - | 2 | 969 | |||
BC039784.1 | - | 2 | 980 | |||
DT929445.1 | - | 4 | 407 |