Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zic4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009576.2 | NP_033602.2 | 4 | 1057 | ||
GenBank mRNA | AK028305.1 | - | 2 | 860 | ||
AK035908.1 | - | 2 | 861 | |||
AK078127.1 | - | 2 | 174 | |||
D78174.1 | - | 2 | 1310 |