Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfbr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011578.3 | NP_035708.2 | 3 | 630 | ||
GenBank mRNA | AF039601.1 | - | 3 | 430 | ||
AK165763.1 | - | 3 | 546 | |||
AK166082.1 | - | 3 | 631 | |||
BC070428.1 | - | 3 | 555 | 6410554 |