Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nufip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013745.5 | NP_038773.1 | 10 | 3156 | ||
GenBank mRNA | AK032324.1 | - | 10 | 3156 |