Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031199.3 | NP_112476.1 | 6 | 1591 | ||
GenBank mRNA | AK162680.1 | - | 6 | 1356 | ||
U65016.1 | - | 6 | 1425 |