Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022536.2 | NP_071981.1 | 3 | 278 | ||
GenBank mRNA | AF071225.1 | - | 3 | 278 | ||
BC061971.1 | - | 3 | 307 | 5623342 | ||
FQ211358.1 | - | 3 | 282 | |||
FQ212031.1 | - | 3 | 307 | |||
FQ219792.1 | - | 3 | 316 | |||
FQ219926.1 | - | 3 | 328 | |||
FQ221020.1 | - | 3 | 331 | |||
FQ221248.1 | - | 3 | 326 | |||
FQ221362.1 | - | 3 | 305 | |||
FQ222696.1 | - | 3 | 440 | |||
FQ223162.1 | - | 3 | 325 | |||
FQ228087.1 | - | 3 | 321 | |||
FQ229174.1 | - | 3 | 328 | |||
FQ229214.1 | - | 3 | 322 | |||
FQ230004.1 | - | 332 | ||||
FQ231656.1 | - | 3 | 321 | |||
FQ232077.1 | - | 3 | 317 | |||
FQ232920.1 | - | 3 | 331 |