Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap2m1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053837.1 | NP_446289.1 | 10 | 1194 | ||
GenBank mRNA | BC087724.1 | - | 1201 | 7320615 | ||
FQ234012.1 | - | 5 | 456 | |||
M23674.1 | - | 10 | 1194 |